(R)-1-(2-Methoxyphenyl)ethylamine ≥95%

Supplier: Ambeed
Danger

Synonyms: (R)-1-(2-Methoxyphenyl)ethanamine

A499454-1G A499454-100MG
77617-136EA 187.46 USD
77617-136 77747-021
(R)-1-(2-Methoxyphenyl)ethylamine ≥95%
(R)-1-(2-Methoxyphenyl)ethylamine

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MDL Number: MFCD00671657
CAS Number: 68285-23-4
UN: 2922
ADR: 8,6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR