Ceftriaxone disodium salt hemiheptahydrate 99+%
Supplier: Ambeed
|
Danger
|
Synonyms:
Ceftriaxone sodium hydrate, Ceftriaxone sodium trihydrate, Ceftriaxone sodium salt hydrate, (6R,7R)-7-((Z)-2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetamido)-3-(((2-methyl-5,6-dioxo-1,2,5,6-tetrahydro-1,2,4-triazin-3-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, disodium salt, hemiheptahydrate
A487798-5G
A487798-1G
A487798-100MG
A487798-250MG
A487798-25G
77602-112EA
32.8
USD
77602-112
77626-976
77880-058
77880-059
77880-060
Ceftriaxone disodium salt hemiheptahydrate 99+%
Ceftriaxone disodium salt hemiheptahydrate
Precaución: for Research Use Only
Formula:
C₁₈H₁₈N₈O₇S₃ |
MDL Number:
MFCD01750405 CAS Number: 104376-79-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...