2-(2-Bromopyridin-4-yl)acetonitrile ≥98%

Supplier: Ambeed
Warning

A499281-250MG A499281-100MG
77893-006EA 60 USD
77893-006 77893-005
2-(2-Bromopyridin-4-yl)acetonitrile ≥98%
2-(2-Bromopyridin-4-yl)acetonitrile

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MDL Number: MFCD11100738
CAS Number: 312325-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR