6-Chloro-4-hydroxy-2-methylquinoline ≥98%

Supplier: Ambeed
Warning

A487657-1G A487657-5G
77901-765EA 49 USD
77901-765 77900-547
6-Chloro-4-hydroxy-2-methylquinoline ≥98%
6-Chloro-4-hydroxy-2-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈ClNO
MDL Number: MFCD00023981
CAS Number: 15644-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR