N-(4-Methoxybenzyl)-1-butanamine ≥97%

Supplier: Ambeed
Warning

A499150-1G A499150-5G
77882-237EA 9 USD
77882-237 77882-238
N-(4-Methoxybenzyl)-1-butanamine ≥97%
N-(4-Methoxybenzyl)-1-butanamine

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO
MDL Number: MFCD01924419
CAS Number: 3910-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR