Maltotetraose ≥97%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4R,5R)-4-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-3,4-Dihydroxy-6-(hydroxymethyl)-5-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal

A487621-100MG A487621-50MG A487621-1G A487621-250MG
77602-108EA 370.03 USD
77602-108 77602-110 77669-180 77669-182
Maltotetraose ≥97%
Maltotetraose

Precaución: for Research Use Only

Formula: C₂₄H₄₂O₂₁
MDL Number: MFCD00010576
CAS Number: 34612-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR