1-Benzyl-1H-imidazole-4-carboxylic acid ≥95%

Supplier: Ambeed
Warning

A498773-1G A498773-250MG
77900-552EA 327 USD
77900-552 77900-532
1-Benzyl-1H-imidazole-4-carboxylic acid ≥95%
1-Benzyl-1H-imidazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O₂
MW: 202.21 g/mol
MDL Number: MFCD07376016
CAS Number: 676372-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR