Dimethyl ethylmalonate ≥95%

Supplier: Ambeed
Warning

A486978-100G A486978-25G A486978-500G
77626-974EA 40.17 USD
77626-974 77880-056 77880-057
Dimethyl ethylmalonate ≥95%
Dimethyl ethylmalonate

Precaución: for Research Use Only

Formula: C₇H₁₂O₄
MDL Number: MFCD00042839
CAS Number: 26717-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR