2-Chloro-6-methoxy-3-(trifluoromethyl)pyridine ≥97%

Supplier: Ambeed
Danger

A498228-100MG A498228-250MG
77891-011EA 91 USD
77891-011 77891-012
2-Chloro-6-methoxy-3-(trifluoromethyl)pyridine ≥97%
2-Chloro-6-methoxy-3-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₅ClF₃NO
MDL Number: MFCD25958510
CAS Number: 136353-03-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR