2,5-Dimethoxy-1,4-benzoquinone ≥98%

Supplier: Ambeed
Warning

A486590-25G A486590-1G
77617-108EA 161.94 USD
77617-108 77747-002
2,5-Dimethoxy-1,4-benzoquinone ≥98%
2,5-Dimethoxy-1,4-benzoquinone

Precaución: for Research Use Only

Formula: C₈H₈O₄
MDL Number: MFCD00189386
CAS Number: 3117-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR