3-Bromo-5-methylbenzyl alcohol ≥98%

Supplier: Ambeed
Warning

Synonyms: (3-Bromo-5-methylphenyl)methanol, 3-Bromo-5-methylbenzyl Alcohol

A497801-1G A497801-250MG A497801-5G
77891-008EA 50 USD
77891-008 77891-009 77891-010
3-Bromo-5-methylbenzyl alcohol ≥98%
3-Bromo-5-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MDL Number: MFCD18204791
CAS Number: 648439-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR