N1,4-Dimethylbenzene-1,2-diamine ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A486306-1G A486306-250MG
77684-292EA 185 USD
77684-292 77684-293
N1,4-Dimethylbenzene-1,2-diamine ≥95%, Ambeed.Inc
N1,4-Dimethylbenzene-1,2-diamine

Precaución: For research use only.

Formula: C₈H₁₂N₂
MW: 136.19 g/mol
MDL Number: MFCD07437799
CAS Number: 39513-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR