Telatinib ≥99%

Supplier: Ambeed
Warning

A485916-1MG A485916-10MG A485916-5MG A485916-100MG A485916-50MG
77602-102EA 19.48 USD
77602-102 77669-168 77669-170 77872-805 77872-806
Telatinib ≥99%
Telatinib

Precaución: for Research Use Only

Formula: C₂₀H₁₆ClN₅O₃
MDL Number: MFCD18251453
CAS Number: 332012-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR