6-Oxa-5-thia-4-azaspiro[2.4]heptane 5,5-dioxide, N-BOC protected ≥98%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl 6-oxa-5-thia-4-azaspiro[2.4]heptane-4-carboxylate 5,5-dioxide

A497108-1G A497108-250MG
77749-017EA 430 USD
77749-017 77749-018
6-Oxa-5-thia-4-azaspiro[2.4]heptane 5,5-dioxide, N-BOC protected ≥98%
6-Oxa-5-thia-4-azaspiro[2.4]heptane 5,5-dioxide, N-BOC protected

Precaución: for Research Use Only

Formula: C₉H₁₅NO₅S
MDL Number: MFCD31925000
CAS Number: 2023006-14-4
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR