5-Bromo-3-chloro-2-fluorobenzonitrile ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A497059-1G A497059-100MG A497059-250MG A497059-5G
77684-318EA 148 USD
77684-318 77893-000 77893-001 77893-002
5-Bromo-3-chloro-2-fluorobenzonitrile ≥95%, Ambeed.Inc
5-Bromo-3-chloro-2-fluorobenzonitrile

Precaución: For research use only.

Formula: C₇H₂BrClFN
MDL Number: MFCD09800811
CAS Number: 1000577-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR