N-Benzylcinchoninium chloride ≥99%

Supplier: Ambeed
Warning

A485474-1G A485474-250MG A485474-5G
77602-100EA 34.85 USD
77602-100 77880-053 77880-054
N-Benzylcinchoninium chloride ≥99%
N-Benzylcinchoninium chloride

Precaución: for Research Use Only

Formula: C₂₆H₂₉ClN₂O
MDL Number: MFCD00082423
CAS Number: 69221-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR