5-Bromo-2-(trifluoromethyl)benzoic acid ≥98%

Supplier: Ambeed
Warning

A485471-1G A485471-25G
77882-218EA 24 USD
77882-218 77882-219
5-Bromo-2-(trifluoromethyl)benzoic acid ≥98%
5-Bromo-2-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O₂
MDL Number: MFCD12026306
CAS Number: 654-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR