Methyl N-Boc-3-Oxopiperidine-4-carboxylate ≥95%

Supplier: Ambeed
Warning

A485182-5G A485182-1G A485182-250MG
77673-264EA 207 USD
77673-264 77882-216 77882-217
Methyl N-Boc-3-Oxopiperidine-4-carboxylate ≥95%
Methyl N-Boc-3-Oxopiperidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₅
MW: 257.28 g/mol
MDL Number: MFCD12198882
CAS Number: 220223-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR