4-(3-Bromophenyl)piperazine, N1-BOC protected ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3-[4-(tert-butoxycarbonyl)piperazin-1-yl]benzene, 4-(3-Bromophenyl)-1-(tert-butoxycarbonyl)piperazine, tert-Butyl 4-(3-bromophenyl)piperazine-1-carboxylate

A496722-1G A496722-5G
77613-574EA 20.6 USD
77613-574 77684-317
4-(3-Bromophenyl)piperazine, N1-BOC protected ≥98%
4-(3-Bromophenyl)piperazine, N1-BOC protected

Precaución: For research use only.

Formula: C₁₅H₂₁BrN₂O₂
MW: 341.243 g/mol
MDL Number: MFCD07781211
CAS Number: 327030-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR