(R)-2-Hydroxypentanedioic acid ≥90%

Supplier: Ambeed
Warning

A496476-100MG A496476-250MG A496476-50MG A496476-1G
77613-564EA 267.8 USD
77613-564 77613-566 77613-568 77686-301
(R)-2-Hydroxypentanedioic acid ≥90%
(R)-2-Hydroxypentanedioic acid

Precaución: for Research Use Only

Formula: C₅H₈O₅
MDL Number: MFCD16251406
CAS Number: 13095-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR