4-(2H-Benzo[d][1,2,3]triazol-2-yl)benzene-1,3-diol 95+%

Supplier: Ambeed
Warning

A496264-1G A496264-5G
77669-226EA 22 USD
77669-226 77872-827
4-(2H-Benzo[d][1,2,3]triazol-2-yl)benzene-1,3-diol 95+%
4-(2H-Benzo[d][1,2,3]triazol-2-yl)benzene-1,3-diol

Precaución: for Research Use Only

Formula: C₁₂H₉N₃O₂
MDL Number: MFCD11870761
CAS Number: 22607-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR