Disodium hydrogen citrate ≥97%

Supplier: Ambeed
Warning

Synonyms: Disodium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate, Sodium 2-(carboxymethyl)-2-hydroxysuccinate, Sodium citrate dibasic, Citric acid disodium salt

A484181-100G A484181-500G
77594-508EA 6.15 USD
77594-508 77880-051
Disodium hydrogen citrate ≥97%
Disodium hydrogen citrate

Precaución: for Research Use Only

Formula: C₆H₆Na₂O₇
MDL Number: MFCD00036367
CAS Number: 144-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR