Fmoc-(S)-3-amino-4-(1-naphthyl)butyric acid ≥98%
Supplier: Ambeed
|
Warning
|
A495144-100MG
A495144-1G
A495144-250MG
77882-229EA
18
USD
77882-229
77882-230
77882-231
Fmoc-(S)-3-amino-4-(1-naphthyl)butyric acid ≥98%
Fmoc-(S)-3-amino-4-(1-naphthyl)butyric acid
Precaución: for Research Use Only
Formula:
C₂₉H₂₅NO₄ |
MDL Number:
MFCD01861034 CAS Number: 270063-38-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...