Ethanesulphonyl chloride ≥98%

Supplier: Ambeed
Danger

A495132-100G A495132-5G A495132-25G
77594-552EA 19.48 USD
77594-552 77602-134 77669-222
Ethanesulphonyl chloride ≥98%
Ethanesulphonyl chloride

Precaución: for Research Use Only

Formula: C₂H₅ClO₂S
MW: 128.58 g/mol
Boiling Pt: 177
Melting Pt: -68
MDL Number: MFCD00007460
CAS Number: 594-44-5
UN: 2927
ADR: 6.1,8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR