5-Methyl-1H-benzo[d]imidazole-2-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A483996-250MG A483996-5G
77602-094EA 53.3 USD
77602-094 77882-211
5-Methyl-1H-benzo[d]imidazole-2-carboxylic acid ≥98%
5-Methyl-1H-benzo[d]imidazole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MDL Number: MFCD03368381
CAS Number: 99459-47-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR