(S)-2-Amino-2-methyl-3-phenylpropanoic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: H-α-Me-Phe-OH

A483861-1G A483861-250MG A483861-5G
77602-092EA 89.18 USD
77602-092 77626-960 77626-962
(S)-2-Amino-2-methyl-3-phenylpropanoic acid ≥95%
(S)-2-Amino-2-methyl-3-phenylpropanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.219 g/mol
MDL Number: MFCD00145244
CAS Number: 23239-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR