3-Fluoro-2-(trifluoromethyl)phenylboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (3-Fluoro-2-(trifluoromethyl)phenyl)boronic acid, 2-(Trifluoromethyl)-3-fluoro-phenylboronic acid

A494842-100MG A494842-1G
77673-288EA 39 USD
77673-288 77872-821
3-Fluoro-2-(trifluoromethyl)phenylboronic acid ≥98%
3-Fluoro-2-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₄O₂
MW: 207.918 g/mol
MDL Number: MFCD08706086
CAS Number: 850411-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR