1-Methyl-3-(trifluoromethyl)-1H-pyrazole-5-boronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)- 3-(trifluoromethyl)-1H-pyrazole, 1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)-1H-pyrazole, 1-METHYL-3-(TRIFLUOROMETHYL)PYRAZOLE-5-BORONIC ACID PINACOL ESTER

A483661-5G A483661-100MG A483661-1G A483661-250MG
77572-528EA 139 USD
77572-528 77673-256 77673-258 77882-210
1-Methyl-3-(trifluoromethyl)-1H-pyrazole-5-boronic acid pinacol ester ≥98%
1-Methyl-3-(trifluoromethyl)-1H-pyrazole-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₆BF₃N₂O₂
MDL Number: MFCD12022323
CAS Number: 1025719-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR