4-Bromo-7-nitroisoindolin-1-one ≥96%

Supplier: Ambeed
Warning

A483352-100MG A483352-250MG
77892-969EA 9 USD
77892-969 77892-970
4-Bromo-7-nitroisoindolin-1-one ≥96%
4-Bromo-7-nitroisoindolin-1-one

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O₃
MDL Number: MFCD18432551
CAS Number: 765948-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR