2,3-O-Isopropylidene-D-erythronolactone ≥98%

Supplier: Ambeed
Warning

Synonyms: 2,3-O-Isopropylidene-D-erythronolactone, (3aR,6aR)-2,2-Dimethyldihydrofuro[3,4-d][1,3]dioxol-4(3aH)-one

A483163-250MG A483163-1G A483163-5G
77617-094EA 25.75 USD
77617-094 77746-994 77746-995
2,3-O-Isopropylidene-D-erythronolactone ≥98%
2,3-O-Isopropylidene-D-erythronolactone

Precaución: for Research Use Only

Formula: C₇H₁₀O₄
MW: 158.15 g/mol
MDL Number: MFCD00134440
CAS Number: 25581-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR