7-Chloro-1,6-naphthyridin-2(1H)-one ≥98%

Supplier: Ambeed
Warning

A483095-1G A483095-250MG
77572-524EA 65 USD
77572-524 77748-999
7-Chloro-1,6-naphthyridin-2(1H)-one ≥98%
7-Chloro-1,6-naphthyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O
MDL Number: MFCD22548048
CAS Number: 1345091-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR