Methyl 4-amino-2,6-dimethoxybenzoate ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A483069-1G A483069-100MG A483069-250MG
77684-284EA 397 USD
77684-284 77890-978 77890-979
Methyl 4-amino-2,6-dimethoxybenzoate ≥98%, Ambeed.Inc
Methyl 4-amino-2,6-dimethoxybenzoate

Precaución: For research use only.

Formula: C₁₀H₁₃NO₄
MDL Number: MFCD18642897
CAS Number: 3956-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR