2-Chloro-3-fluoro-5-pyridineboronic acid ≥95%

Supplier: Ambeed
Warning

A493897-1G A493897-250MG A493897-5G A493897-25G
77594-548EA 45.1 USD
77594-548 77669-216 77669-218 77872-820
2-Chloro-3-fluoro-5-pyridineboronic acid ≥95%
2-Chloro-3-fluoro-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₄BClFNO₂
MDL Number: MFCD04112500
CAS Number: 1072946-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR