2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)furan-2-carboxylate

A493708-1G A493708-250MG A493708-5G
77886-280EA 143 USD
77886-280 77886-281 77886-282
2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester ≥98%
2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₉BO₅
MDL Number: MFCD11855980
CAS Number: 1073338-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR