2-Bromo-3-methoxyisonicotinaldehyde ≥95%

Supplier: Ambeed
Danger

A479105-250MG A479105-100MG
77673-238EA 119 USD
77673-238 77882-203
2-Bromo-3-methoxyisonicotinaldehyde ≥95%
2-Bromo-3-methoxyisonicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MDL Number: MFCD06410674
CAS Number: 191418-78-7
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR