(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-(tert-butoxy)propanoic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-N-Me-Ser(tBu)-OH

A492305-1G A492305-250MG A492305-5G A492305-100MG
77892-996EA 160 USD
77892-996 77892-997 77892-998 77892-995
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-(tert-butoxy)propanoic acid ≥95%
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-(tert-butoxy)propanoic acid

Precaución: for Research Use Only

Formula: C₂₃H₂₇NO₅
MW: 397.46 g/mol
MDL Number: MFCD28656714
CAS Number: 1210833-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR