(2-(2-(dimethylamino)ethoxy)phenyl)boronic acid ≥95%

Supplier: Ambeed
Warning

A492080-100MG A492080-1G A492080-250MG
77892-992EA 131 USD
77892-992 77892-993 77892-994
(2-(2-(dimethylamino)ethoxy)phenyl)boronic acid ≥95%
(2-(2-(dimethylamino)ethoxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₆BNO₃
MDL Number: MFCD11183160
CAS Number: 1313760-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR