Galloflavin ≥98%

Supplier: Ambeed
Warning

A477947-100MG A477947-250MG
77890-962EA 287 USD
77890-962 77890-963
Galloflavin ≥98%
Galloflavin

Precaución: for Research Use Only

Formula: C₁₂H₆O₈
MDL Number: MFCD26941292
CAS Number: 568-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR