1-(1-methylazetidin-3-yl)methanamine dihydrochloride ≥98%

Supplier: Ambeed
Warning

A477826-250MG A477826-1G A477826-5G
77594-492EA 71.75 USD
77594-492 77622-332 77783-432
1-(1-methylazetidin-3-yl)methanamine dihydrochloride ≥98%
1-(1-methylazetidin-3-yl)methanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₄Cl₂N₂
MDL Number: MFCD28125097
CAS Number: 1803609-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR