iCRT14 ≥98%

Supplier: Ambeed
Warning

Synonyms: 5-((2,5-Dimethyl-1-(pyridin-3-yl)-1H-pyrrol-3-yl)methylene)-3-phenylthiazolidine-2,4-dione, iCRT 14

A491892-10MG A491892-5MG
77686-287EA 75 USD
77686-287 77686-288
iCRT14 ≥98%
iCRT14

Precaución: for Research Use Only

Formula: C₂₁H₁₇N₃O₂S
MW: 375.45 g/mol
MDL Number: MFCD04366833
CAS Number: 677331-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR