Phenyl α-D-thiomannopyranoside ≥97%

Supplier: Ambeed
Warning

Synonyms: Phenyl 1-thio-α-D-mannopyranoside, (2R,3S,4S,5S,6R)-2-(Hydroxymethyl)-6-(phenylthio)tetrahydro-2H-pyran-3,4,5-triol

A477633-250MG A477633-100MG A477633-1G
77617-070EA 96.82 USD
77617-070 77746-982 77746-983
Phenyl α-D-thiomannopyranoside ≥97%
Phenyl α-D-thiomannopyranoside

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₅S
MW: 272.32 g/mol
MDL Number: MFCD06657857
CAS Number: 77481-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR