N4-Benzoyl-2'-O-methylcytidine ≥98%

Supplier: Ambeed
Warning

Synonyms: N-(1-((2R,3R,4R,5R)-4-Hydroxy-5-(hydroxymethyl)-3-methoxytetrahydrofuran-2-yl)-2-oxo-1,2-dihydropyrimidin-4-yl)benzamide, N4-Benzoyl-2’-O-methylcytidine

A491606-100MG A491606-250MG
77594-538EA 13.33 USD
77594-538 77602-122
N4-Benzoyl-2'-O-methylcytidine ≥98%
N4-Benzoyl-2'-O-methylcytidine

Precaución: for Research Use Only

Formula: C₁₇H₁₉N₃O₆
MW: 361.35 g/mol
MDL Number: MFCD01631038
CAS Number: 52571-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR