5-Methoxybenzo[b]thiophene-2-carbaldehyde ≥98%

Supplier: Ambeed
Warning

A476980-1G A476980-250MG
77613-492EA 170.98 USD
77613-492 77673-230
5-Methoxybenzo[b]thiophene-2-carbaldehyde ≥98%
5-Methoxybenzo[b]thiophene-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₈O₂S
MW: 192.234 g/mol
MDL Number: MFCD11101036
CAS Number: 622864-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR