Fmoc-(S)-3-amino-4-(4-cyanophenyl)butyric acid ≥95%

Supplier: Ambeed
Warning

A482551-250MG A482551-100MG
77673-250EA 76 USD
77673-250 77882-209
Fmoc-(S)-3-amino-4-(4-cyanophenyl)butyric acid ≥95%
Fmoc-(S)-3-amino-4-(4-cyanophenyl)butyric acid

Precaución: for Research Use Only

Formula: C₂₆H₂₂N₂O₄
MDL Number: MFCD01861080
CAS Number: 270065-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR