5-Fluoroquinoline-8-carboxylic acid ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A482241-250MG A482241-100MG A482241-1G
77684-280EA 178 USD
77684-280 77890-975 77890-976
5-Fluoroquinoline-8-carboxylic acid ≥98%, Ambeed.Inc
5-Fluoroquinoline-8-carboxylic acid

Precaución: For research use only.

Formula: C₁₀H₆FNO₂
MDL Number: MFCD09048900
CAS Number: 926252-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR