(2-Fluoro-5-nitrophenyl)acetic acid ≥98%

Supplier: Ambeed
Warning

A482110-5G A482110-25G
77669-162EA 18 USD
77669-162 77872-797
(2-Fluoro-5-nitrophenyl)acetic acid ≥98%
(2-Fluoro-5-nitrophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₆FNO₄
MDL Number: MFCD04091773
CAS Number: 195609-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR