Chloro(crotyl)[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II) ≥98%

Supplier: Ambeed
Warning

A476171-1G A476171-250MG
77783-430EA 261 USD
77783-430 77783-431
Chloro(crotyl)[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II) ≥98%
Chloro(crotyl)[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II)

Precaución: for Research Use Only

Formula: C₂₀H₃₅ClNPPd
MDL Number: MFCD29059853
CAS Number: 1334497-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR