Diethyl disulfide ≥98%

Supplier: Ambeed
Danger

A476161-25G A476161-100G
77602-068EA 32.8 USD
77602-068 77880-034
Diethyl disulfide ≥98%
Diethyl disulphide

Precaución: for Research Use Only

Formula: C₄H₁₀S₂
MDL Number: MFCD00009266
CAS Number: 110-81-6
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR