2-{[2-(4-Chlorophenoxy)ethyl]thio}-1H-benzimidazole ≥97%

Supplier: Ambeed
Warning

Synonyms: CLP-3094, 2-((2-(4-Chlorophenoxy)ethyl)thio)-1H-benzo[d]imidazole

A475953-50MG A475953-100MG
77746-977EA 98 USD
77746-977 77746-976
2-{[2-(4-Chlorophenoxy)ethyl]thio}-1H-benzimidazole ≥97%
2-{[2-(4-Chlorophenoxy)ethyl]thio}-1H-benzimidazole

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClN₂OS
MW: 304.8 g/mol
MDL Number: MFCD00805733
CAS Number: 312749-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR