propargyl-PEG5-Acid ≥95%

Supplier: Ambeed
Warning

Synonyms: 4,7,10,13,16-Pentaoxanonadec-18-yn-1-oic acid

A481703-1G A481703-250MG
77572-516EA 57.68 USD
77572-516 77617-086
propargyl-PEG5-Acid ≥95%
propargyl-PEG5-Acid

Precaución: for Research Use Only

Formula: C₁₄H₂₄O₇
MW: 304.336 g/mol
MDL Number: MFCD22201536
CAS Number: 1245823-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR